Läs mer
3-Chloro-L-tyrosine, TRC
Produktkod. 30138489
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138489 1 g
30005260 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138489 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4231001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7423-93-0
Molekylformel C9 H10 N O3 Cl
Molekylvikt (g/mol) 215.63
Synonym meta-Chlorotyrosine
IUPAC-namn (2S)-2-amino-3-(3-chloro-4-hydroxy-phenyl)propanoic acid
LEDER N[C@@H](Cc1ccc(O)c(Cl)c1)C(=O)O
Specifikationer
Kemiskt namn eller material 3-Chloro-L-tyrosine
InChI formel InChI=1S/C9H10ClNO3/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14/h1-3,7,12H,4,11H2,(H,13,14)/t7-/m0/s1
Kvantitet 1 g
Formel vikt 215.0349