Learn More
3-Chloro-N-methylpropan-1-amine Hydrochloride, TRC
Product Code. 30351992
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30351992 1 g 1g
1 options
Product Code. 30351992 Supplier Toronto Research Chemicals Supplier No. TRCC3805981G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloro-N-methylpropan-1-amine Hydrochloride
CAS 97145-88-5
Molecular Formula C4H10ClN•(HCl)
Quantity 1 g
Synonym Fluoxetine Impurity, 3-(Methylamino)propyl Chloride Hydrochloride, N-Methyl(3-chloropropyl)amine Hydrochloride
InChI Formula InChI=1S/C4H10ClN.ClH/c1-6-4-2-3-5;/h6H,2-4H2,1H3;1H
SMILES CNCCCCl.Cl
IUPAC Name 3-chloro-N-methylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 107.583646
Formula Weight 143.0269