Läs mer
3-Chloroacenaphthene, TRC
Produktkod. 30367414
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367414 10 mg
30350001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367414 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC35340010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Chloroacenaphthene
CAS 5573-31-9
Molekylformel C12H9Cl
Kvantitet 10 mg
Synonym 3-Chloro-1,2-dihydroacenaphthylene
InChI formel InChI=1S/C12H9Cl/c13-11-7-5-9-3-1-2-8-4-6-10(11)12(8)9/h1-3,5,7H,4,6H2
LEDER Clc1ccc2cccc3CCc1c23
IUPAC-namn 3-chloro-1,2-dihydroacenaphthylene
Molekylvikt (g/mol) 188.65
Formel vikt 188.0393