Läs mer
3-Chlorocetirizine Dihydrochloride ((RS)-2-[2-[4-[(3-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride), Mikromol™
Produktkod. 17971486
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17971486 25 mg
17941806 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17971486 Leverantör Mikromol Leverantörsnummer MM0380.170025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Chlorocetirizine Dihydrochloride ((RS)-2-[2-[4-[(3-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Chlorocetirizine Dihydrochloride
CAS 1232460-29-5
Analyt- eller komponentnamn 3-Chlorocetirizine Dihydrochloride
Frakt skick Room Temperature
Molekylformel C21 H25 Cl N2 O3 . 2 Cl H
Certifikat och efterlevnad ISO 9001
Synonym 3-Chlorocetirizine Dihydrochloride, (RS)-2-[2-[4-[(3-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride
InChI formel InChI=1S/C21H25ClN2O3.2ClH/c22-19-8-4-7-18(15-19)21(17-5-2-1-3-6-17)24-11-9-23(10-12-24)13-14-27-16-20(25)26;;/h1-8,15,21H,9-14,16H2,(H,25,26);2*1H
LEDER Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3cccc(Cl)c3
IUPAC-namn 2-[2-[4-[(3-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
Molekylvikt (g/mol) 461.81
Kvantitet 25 mg
Formel vikt 460.1087 g/mol