Läs mer
3-Chlorocyclohexene, TRC
Produktkod. 30350398
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350398 50 mg
30361313 100 mg
30354510 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30350398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38425550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2441-97-6
Molekylformel C6H9Cl
Molekylvikt (g/mol) 116.59
Synonym Cyclohexene, 3-chloro- (6CI, 8CI, 9CI, ACI), 3-Chlorocyclohexene (ACI), 1-Chloro-2-cyclohexene, 2-Cyclohexenyl chloride, 3-Chloro-1-cyclohexene, 3-Cyclohexenyl chloride
IUPAC-namn 3-chlorocyclohexene
LEDER ClC1CCCC=C1
Specifikationer
Kemiskt namn eller material 3-Chlorocyclohexene
Kvantitet 50 mg
InChI formel InChI=1S/C6H9Cl/c7-6-4-2-1-3-5-6/h2,4,6H,1,3,5H2
Formel vikt 116.0393