Läs mer
3-Chloroindomethacin, TRC
Produktkod. 30012494
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012494 50 mg
30004532 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36739550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 402849-26-7
Molekylformel C19 H15 Cl2 N O4
Molekylvikt (g/mol) 392.23
Synonym 1H-Indole-3-acetic acid, 1-(3,4-dichlorobenzoyl)-5-methoxy-2-methyl-, 1-(3,4-Dichlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid, [1-(3,4-Dichlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid, 3,4-Dichloroindometacin, [1-(3,4-Dichlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid, 3-Chloroindomethacin
IUPAC-namn 2-[1-(3,4-dichlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
LEDER COc1ccc2c(c1)c(CC(=O)O)c(C)n2C(=O)c3ccc(Cl)c(Cl)c3
Specifikationer
Kemiskt namn eller material (1-(3,4-Dichlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetic Acid (3,4-Dichloroindometacin)
Kvantitet 50 mg
InChI formel InChI=1S/C19H15Cl2NO4/c1-10-13(9-18(23)24)14-8-12(26-2)4-6-17(14)22(10)19(25)11-3-5-15(20)16(21)7-11/h3-8H,9H2,1-2H3,(H,23,24)
Formel vikt 391.0378