Läs mer
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole Hydrochloride, TRC
Produktkod. 30344989
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30344989 100 mg
1 options
Produktkod. 30344989 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB421138100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole Hydrochloride
CAS 135206-76-7
Molekylformel C4H6N3Cl.HCl
Kvantitet 100 mg
Synonym 1H-1,2,4-Triazole, 3-(chloromethyl)-1-methyl-, hydrochloride (1:1) (ACI), 1H-1,2,4-Triazole, 3-(chloromethyl)-1-methyl-, monohydrochloride (9CI), 3-Chloromethyl-1-methyl-1H-[1,2,4]triazole hydrochloride, 3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride
InChI formel InChI=1S/C4H6ClN3.ClH/c1-8-3-6-4(2-5)7-8;/h3H,2H2,1H3;1H
LEDER Cl.Cn1cnc(CCl)n1
IUPAC-namn 3-(chloromethyl)-1-methyl-1,2,4-triazole;hydrochloride
Molekylvikt (g/mol) 168.02
Formel vikt 167.0017