missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1-methyl-1H-indazole, ≥97%, Thermo Scientific™

Catalog Number p-3174969
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1578-97-8
C9H9ClN2
180.64
MFCD08060497
LYLGQIOTMAQQEB-UHFFFAOYSA-N
3-chloromethyl-1-methyl-1h-indazole, 3-chloromethyl-1-methylindazole, 1h-indazole, 3-chloromethyl-1-methyl, 1-methyl-3-chloromethylindazole, 1h-indazole,3-chloromethyl-1-methyl
7537470
3-(chloromethyl)-1-methylindazole
CN1N=C(CCl)C2=CC=CC=C12
This item is not returnable. View return policy
1578-97-8
C9H9ClN2
180.64
MFCD08060497
LYLGQIOTMAQQEB-UHFFFAOYSA-N
3-chloromethyl-1-methyl-1h-indazole, 3-chloromethyl-1-methylindazole, 1h-indazole, 3-chloromethyl-1-methyl, 1-methyl-3-chloromethylindazole, 1h-indazole,3-chloromethyl-1-methyl
7537470
3-(chloromethyl)-1-methylindazole
CN1N=C(CCl)C2=CC=CC=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.64
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-1-methyl-1H-indazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'