missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-(2-furyl)-1-methyl-1H-pyrazole, 95%, Thermo Scientific™

Catalog Number p-3485344
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
876316-48-2
C9H9ClN2O
196.63
MFCD08690269
DZUIHVJDKDNADW-UHFFFAOYSA-N
3-chloromethyl-5-2-furyl-1-methyl-1h-pyrazole, 3-chloromethyl-5-furan-2-yl-1-methylpyrazole, 1h-pyrazole,3-chloromethyl-5-2-furanyl-1-methyl, 3-chloromethyl-5-furan-2-yl-1-methyl-1h-pyrazole, 2-3-chloromethyl-1-methylpyrazol-5-yl furan
18525803
3-(chloromethyl)-5-(furan-2-yl)-1-methylpyrazole
CN1N=C(CCl)C=C1C1=CC=CO1
This item is not returnable. View return policy
876316-48-2
C9H9ClN2O
196.63
MFCD08690269
DZUIHVJDKDNADW-UHFFFAOYSA-N
3-chloromethyl-5-2-furyl-1-methyl-1h-pyrazole, 3-chloromethyl-5-furan-2-yl-1-methylpyrazole, 1h-pyrazole,3-chloromethyl-5-2-furanyl-1-methyl, 3-chloromethyl-5-furan-2-yl-1-methyl-1h-pyrazole, 2-3-chloromethyl-1-methylpyrazol-5-yl furan
18525803
3-(chloromethyl)-5-(furan-2-yl)-1-methylpyrazole
CN1N=C(CCl)C=C1C1=CC=CO1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 196.64
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-(2-furyl)-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'