missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Chloropropiophenone, 98%, Thermo Scientific Chemicals
291.12 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3'-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 45°C to 47°C |
Boiling Point | 123°C to 124°C (13 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
Solubility Information | Soluble in methanol. |
Formula Weight | 168.62 |
Percent Purity | 98% |
Chemical Name or Material | 3'-Chloropropiophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'