Läs mer
3-Chloropyridazine, TRC
Produktkod. 30349722
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349722 1 g
30362904 500 mg
30355974 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349722 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5962931G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1120-95-2
Molekylformel C4H3N2Cl
Molekylvikt (g/mol) 114.53
Synonym Pyridazine, 3-chloro- (6CI, 7CI, 8CI, 9CI, ACI), 3-Chloropyridazine (ACI), 6-Chloropyridazine
IUPAC-namn 3-chloropyridazine
LEDER Clc1cccnn1
Specifikationer
Kemiskt namn eller material 3-Chloropyridazine
Kvantitet 1 g
InChI formel InChI=1S/C4H3ClN2/c5-4-2-1-3-6-7-4/h1-3H
Formel vikt 113.9985