Läs mer
3-CPMT Hydrochloride, TRC
Produktkod. 30356434
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30356434 25 mg
1 options
Produktkod. 30356434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC78123325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-CPMT HCl
CAS 14008-79-8
Molekylformel C21 H24 Cl N O . H Cl
Kvantitet 25 mg
Synonym Chlortropbenzyl, FC 1, SL 6057, Tropine 4-Chlorobenzhydryl Ether Hydrochloride, WY 2149, endo-3-[(4-Chlorophenyl)phenylmethoxy]-8-methyl-8-azabicyclo[3.2.1]octane Hydrochloride, (3-endo)-3-[(4-Chlorophenyl)phenylmethoxy]-8-methyl-8-azabicyclo[3.2.1]octane Hydrochloride
InChI formel InChI=1S/C21H24ClNO.ClH/c1-23-18-11-12-19(23)14-20(13-18)24-21(15-5-3-2-4-6-15)16-7-9-17(22)10-8-16;/h2-10,18-21H,11-14H2,1H3;1H/t18-,19+,20+,21?;
LEDER Cl.CN1[C@H]2CC[C@@H]1CC(C2)OC(c3ccccc3)c4ccc(Cl)cc4
IUPAC-namn (1S,5R)-3-[(4-chlorophenyl)-phenyl-methoxy]-8-methyl-8-azabicyclo[3.2.1]octane;hydrochloride
Molekylvikt (g/mol) 378.34
Formel vikt 377.1313