missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
3-Cyano-2-ethyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea, Mikromol™
Beskrivning
3-Cyano-2-ethyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-Cyano-2-ethyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isourea |
| CAS | 75850-09-8 |
| Analyt- eller komponentnamn | 3-Cyano-2-ethyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isourea |
| Frakt skick | Room Temperature |
| Molekylformel | C11 H17 N5 O S |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Methylethyl Analogue of Oxybutynin Hydrochloride, N-Cyano-N'-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]carbamimidic Acid Ethyl Ester (9CI), 3-Cyano-2-ethyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea |
| InChI formel | InChI=1S/C11H17N5OS/c1-3-17-11(14-7-12)13-4-5-18-6-10-9(2)15-8-16-10/h8H,3-6H2,1-2H3,(H,13,14)(H,15,16) |
| LEDER | CCOC(=NC#N)NCCSCc1nc[nH]c1C |
| IUPAC-namn | ethyl N'-cyano-N-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]carbamimidate |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?