Läs mer
3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea, Mikromol™
Produktkod. 30052543
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052543 100 mg
30051043 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052543 Leverantör Mikromol Leverantörsnummer MM0020.12
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
CAS 138035-55-9
Analyt- eller komponentnamn 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
Frakt skick Room Temperature
Molekylformel C10 H15 N5 O S
Certifikat och efterlevnad ISO 17025
Synonym Cimetidine Hydrochloride Imp. B (EP), Cimetidine Imp. B (EP), 3-Cyano-2-methyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea, Cimetidine Hydrochloride Impurity B, Cimetidine Impurity B, Cimetidine Impurity B
InChI formel InChI=1S/C10H15N5OS/c1-8-9(15-7-14-8)5-17-4-3-12-10(16-2)13-6-11/h7H,3-5H2,1-2H3,(H,12,13)(H,14,15)
LEDER COC(=NC#N)NCCSCc1nc[nH]c1C
IUPAC-namn methyl N'-cyano-N-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]carbamimidate
Molekylvikt (g/mol) 253.32
Kvantitet 100 mg
Formel vikt 253.0997 g/mol