Läs mer
3-Cyano-6,7-dihydro-4-trifluoromethyl-1,5H-cyclopenta-[b]-pyrid-2-one, TRC
Produktkod. 30356509
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356509 100 mg
30354295 50 mg
30355685 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC999263100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 371967-27-0
Molekylformel C10H7N2OF3
Molekylvikt (g/mol) 228.17
IUPAC-namn 2-oxo-4-(trifluoromethyl)-1,5,6,7-tetrahydrocyclopenta[b]pyridine-3-carbonitrile
LEDER Oc1nc2CCCc2c(c1C#N)C(F)(F)F
Specifikationer
Kemiskt namn eller material 3-Cyano-6,7-dihydro-4-trifluoromethyl-1,5H-cyclopenta-[b]-pyrid-2-one
Kvantitet 100 mg
InChI formel InChI=1S/C10H7F3N2O/c11-10(12,13)8-5-2-1-3-7(5)15-9(16)6(8)4-14/h1-3H2,(H,15,16)
Formel vikt 228.051