Läs mer
3-(Cyanomethyl)benzoic Acid, TRC
Produktkod. 30353009
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30353009 250 mg
1 options
Produktkod. 30353009 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC981890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(Cyanomethyl)benzoic Acid
CAS 5689-33-8
Molekylformel C9 H7 N O2
Kvantitet 250 mg
Synonym 3-(Cyanomethyl)benzoic acid, Ketoprofen Imp. H (EP), Benzoic acid, 3-(cyanomethyl)-, m-Toluic acid, α-cyano- (6CI,7CI,8CI), 3-Carboxyphenylacetonitrile, m-(Cyanomethyl)benzoic acid
InChI formel InChI=1S/C9H7NO2/c10-5-4-7-2-1-3-8(6-7)9(11)12/h1-3,6H,4H2,(H,11,12)
LEDER OC(=O)c1cccc(CC#N)c1
IUPAC-namn 3-(cyanomethyl)benzoic acid
Molekylvikt (g/mol) 161.16
Formel vikt 161.0477