Learn More
Alfa Aesar™ 3-[Cyclohexyl(methyl)carbamoyl]-2-fluorobenzeneboronic acid, 97%
Brand:  Alfa Aesar™ H56223.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15482556
  • 5881.31 SEK / 1g
Estimated Shipment date
from Supplier 01-02-2018
Add to basket
Specifications
Chemical Identifiers
3-[Cyclohexyl(methyl)carbamoyl]-2-fluorobenzeneboronic acid
279.11
MFCD22682852
Specifications
3-[Cyclohexyl(methyl)carbamoyl]-2-fluorobenzeneboronic acid
Solid
279.11
1g
97%
MFCD22682852