Läs mer
3-Cyclohexylbutanoic Acid, TRC
Produktkod. 30366950
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366950 250 mg
30368701 50 mg
30361743 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC993708250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4361-38-0
Molekylformel C10H18O2
Molekylvikt (g/mol) 170.25
IUPAC-namn 3-cyclohexylbutanoic acid
LEDER CC(CC(=O)O)C1CCCCC1
Specifikationer
Kemiskt namn eller material 3-Cyclohexylbutanoic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C10H18O2/c1-8(7-10(11)12)9-5-3-2-4-6-9/h8-9H,2-7H2,1H3,(H,11,12)
Formel vikt 170.1307