Läs mer
3-Cyclopropyl-4-methyl-1h-pyrazole-5-carboxylic Acid (>80%), TRC
Produktkod. 30367033
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367033 100 mg
30353522 500 mg
30364044 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30367033 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC230576100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1867157-28-5
Molekylformel C8H10N2O2
Molekylvikt (g/mol) 166.18
Synonym 5-Cyclopropyl-4-methyl-1H-pyrazole-3-carboxylic Acid
IUPAC-namn 5-cyclopropyl-4-methyl-1H-pyrazole-3-carboxylic acid
LEDER Cc1c([nH]nc1C(=O)O)C2CC2
Specifikationer
Kemiskt namn eller material 3-Cyclopropyl-4-methyl-1h-pyrazole-5-carboxylic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C8H10N2O2/c1-4-6(5-2-3-5)9-10-7(4)8(11)12/h5H,2-3H2,1H3,(H,9,10)(H,11,12)
Formel vikt 166.0742