Läs mer
3-Cyclopropyl Hydroxychloroquine, TRC
Produktkod. 30366486
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366486 25 mg
30350875 5 mg
30354946 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366486 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98946125MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2054859-10-6
Molekylformel C21H30ClN3O
Molekylvikt (g/mol) 375.94
Synonym 2-((4-((7-Chloro-3-cyclopropylquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol
IUPAC-namn 2-[4-[(7-chloro-3-cyclopropylquinolin-4-yl)amino]pentyl-ethylamino]ethanol
LEDER CCN(CCO)CCCC(NC1=C2C=CC(Cl)=CC2=NC=C1C3CC3)C
Specifikationer
Kemiskt namn eller material 3-Cyclopropyl Hydroxychloroquine
Kvantitet 25 mg
InChI formel InChI=1S/C21H30ClN3O/c1-3-25(11-12-26)10-4-5-15(2)24-21-18-9-8-17(22)13-20(18)23-14-19(21)16-6-7-16/h8-9,13-16,26H,3-7,10-12H2,1-2H3,(H,23,24)