missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15498696
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
162401-62-9
C12H12F2O4
258.221
MFCD04621687
IGFDIFLMMLWKKY-UHFFFAOYSA-N
3-cyclopropylmethoxy-4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy, roflumilast intermediate, roflumilast intermediate b, 3-cyclopropylmethoxy-4-difluoromethoxybenzoicacid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, 3-cyclopropylmethoxy 4-difluoromethoxy benzoic acid
2394006
3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
C1CC1COC2=C(C=CC(=C2)C(=O)O)OC(F)F
This item is not returnable. View return policy
162401-62-9
C12H12F2O4
258.221
MFCD04621687
IGFDIFLMMLWKKY-UHFFFAOYSA-N
3-cyclopropylmethoxy-4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy, roflumilast intermediate, roflumilast intermediate b, 3-cyclopropylmethoxy-4-difluoromethoxybenzoicacid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, 3-cyclopropylmethoxy 4-difluoromethoxy benzoic acid
2394006
3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
C1CC1COC2=C(C=CC(=C2)C(=O)O)OC(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 120°C
Quantity 250 mg
Beilstein 11413145
Formula Weight 258.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'