Läs mer
3-Deaza Adenosine, TRC
Produktkod. 30364304
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364304 5 mg
30366437 10 mg
30353079 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2008755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6736-58-9
Molekylformel C11 H14 N4 O4
Molekylvikt (g/mol) 266.25
IUPAC-namn (2R,3R,4S,5R)-2-(4-aminoimidazo[4,5-c]pyridin-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER Nc1nccc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
Specifikationer
Kemiskt namn eller material 3-Deaza Adenosine
Kvantitet 5 mg
InChI formel InChI=1S/C11H14N4O4/c12-10-7-5(1-2-13-10)15(4-14-7)11-9(18)8(17)6(3-16)19-11/h1-2,4,6,8-9,11,16-18H,3H2,(H2,12,13)/t6-,8-,9-,11-/m1/s1
Formel vikt 266.1015