Läs mer
(-)-3-Dehydro Shikimic Acid, TRC
Produktkod. 30135164
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135164 25 mg
30133565 100 mg
30003612 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135164 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23047525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2922-42-1
Molekylformel C7 H8 O5
Molekylvikt (g/mol) 172.14
IUPAC-namn (4S,5R)-4,5-dihydroxy-3-oxocyclohexene-1-carboxylic acid
LEDER O[C@@H]1CC(=CC(=O)[C@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material (-)-3-Dehydroshikimic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C7H8O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1,5-6,9-10H,2H2,(H,11,12)/t5-,6-/m1/s1
Formel vikt 172.0372
Procent renhet >95%