Läs mer
3-Dehydroxy 2,3-Dehydro Atorvastatin Lactone, TRC
Produktkod. 30354030
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30354030 100 mg
1 options
Produktkod. 30354030 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229270100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-[2-[(2S)-3,6-Dihydro-6-oxo-2H-pyran-2-yl]ethyl]-5-(4-fluorophenyl)-2-(1-methylethyl)-N,4-diphenyl-1H-pyrrole-3-carboxamide (Atorvastatin eliminated Lactone)
CAS 442851-50-5
Molekylformel C33 H31 F N2 O3
Kvantitet 100 mg
Synonym 1-[2-[(2S)-3,6-Dihydro-6-oxo-2H-pyran-2-yl]ethyl]-5-(4-fluorophenyl)-2-(1-methylethyl)-N,4-diphenyl-1H-pyrrole-3-carboxamide, Atorvastatin eliminated Lactone
InChI formel InChI=1S/C33H31FN2O3/c1-22(2)31-30(33(38)35-26-12-7-4-8-13-26)29(23-10-5-3-6-11-23)32(24-16-18-25(34)19-17-24)36(31)21-20-27-14-9-15-28(37)39-27/h3-13,15-19,22,27H,14,20-21H2,1-2H3,(H,35,38)/t27-/m0/s1
LEDER CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H]5CC=CC(=O)O5
IUPAC-namn 5-(4-fluorophenyl)-1-[2-[(2S)-6-oxo-2,3-dihydropyran-2-yl]ethyl]-N,4-diphenyl-2-propan-2-ylpyrrole-3-carboxamide
Molekylvikt (g/mol) 522.61
Formel vikt 522.2319