Läs mer
3-Deoxy-1,2,4,6-tetra-O-acetyl-D-glucopyranose, TRC
Produktkod. 30349929
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349929 250 mg
30363037 50 mg
30356909 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349929 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD278775250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5040-09-5
Molekylformel C14 H20 O9
Molekylvikt (g/mol) 332.3
IUPAC-namn [(2R,3S,5R)-3,5,6-triacetyloxyoxan-2-yl]methyl acetate
LEDER CC(=O)OC[C@H]1OC(OC(=O)C)[C@@H](C[C@@H]1OC(=O)C)OC(=O)C
Specifikationer
Kemiskt namn eller material 3-Deoxy-1,2,4,6-tetra-O-acetyl-D-glucopyranose
Kvantitet 250 mg
InChI formel InChI=1S/C14H20O9/c1-7(15)19-6-13-11(20-8(2)16)5-12(21-9(3)17)14(23-13)22-10(4)18/h11-14H,5-6H2,1-4H3/t11-,12+,13+,14?/m0/s1
Formel vikt 332.1107