Learn More
3'-Deoxy-2',3'-didehydrothymidine, 98%, Thermo Scientific Chemicals
Catalog Number p-7077523
Click to view available options
Quantity:
1 g
5 g
3056-17-5
C10H12N2O4
224.22
MFCD00132921
XNKLLVCARDGLGL-HGXVMFPFNA-N
stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine
18283
CHEBI:63581
1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O
3056-17-5
C10H12N2O4
224.22
MFCD00132921
XNKLLVCARDGLGL-HGXVMFPFNA-N
stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine
18283
CHEBI:63581
1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 162.0°C to 169.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 158929
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 3'-Deoxy-2',3'-didehydrothymidine
compliance-icons compliance-icons
Product Identifier
  • 3'-Deoxy-2',3'-didehydrothymidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.