Läs mer
3’-Deoxy-3’-fluoro Thymidine, TRC
Produktkod. 30359239
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359239 250 mg
30361821 50 mg
30357911 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359239 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD235475250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 25526-93-6
Molekylformel C10H13FN2O4
Molekylvikt (g/mol) 244.22
LEDER CC1=CN([C@H]2C[C@H](F)[C@@H](CO)O2)C(=O)NC1=O
Specifikationer
Kemiskt namn eller material 3'-Deoxy-3'-fluoro Thymidine
Kvantitet 250 mg
InChI formel InChI=1S/C10H13FN2O4/c1-5-3-13(10(16)12-9(5)15)8-2-6(11)7(4-14)17-8/h3,6-8,14H,2,4H2,1H3,(H,12,15,16)/t6-,7+,8+/m0/s1