Läs mer
3’-Deoxy-3’-fluorothymidine-5’-monophosphate Disodium Salt, TRC
Produktkod. 30363570
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30363570 1 mg
1 options
Produktkod. 30363570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2354801MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3'-Deoxy-3'-fluorothymidine-5'-monophosphate Disodium Salt
CAS 1316845-74-5
Molekylformel C10H12FN2Na2O7P
Kvantitet 1 mg
Synonym 3'-Deoxy-3'-fluoro-5'-thymidylic Acid Disodium, 2',3'-Dideoxy-3'-fluoro-5- methyluridine 5'-(Dihydrogen Phosphate) Disodium, 3'-Deoxy-3'-fluorothymidine Monophosphate Disodium, 3'-Fluoro-3'-deoxythymidine Monophosphate Disodium
InChI formel InChI=1S/C10H14FN2O7P.2Na/c1-5-3-13(10(15)12-9(5)14)8-2-6(11)7(20-8)4-19-21(16,17)18;;/h3,6-8H,2,4H2,1H3,(H,12,14,15)(H2,16,17,18);;/q;2*+1/p-2/t6-,7+,8+;;/m0../s1
LEDER [Na+].[Na+].CC1=CN([C@H]2C[C@H](F)[C@@H](COP(=O)([O-])[O-])O2)C(=O)NC1=O
Molekylvikt (g/mol) 368.16