missing translation for 'onlineSavingsMsg'
Läs mer

3-Deoxy-D-erythro-hexos-2-ulose-bis-benzoylhydrazone, TRC

Produktkod. 30350719
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30350719 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350719 Supplier Toronto Research Chemicals Supplier No. TRCD2329001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Deoxy-D-erythro-hexos-2-ulose-bis-benzoylhydrazone
CAS 32443-70-2
Molekylformel C20 H22 N4 O5
Kvantitet 1 g
InChI formel InChI=1S/C20H22N4O5/c25-13-18(27)17(26)11-16(22-24-20(29)15-9-5-2-6-10-15)12-21-23-19(28)14-7-3-1-4-8-14/h1-10,12,17-18,25-27H,11,13H2,(H,23,28)(H,24,29)/b21-12+,22-16-/t17-,18+/m0/s1
LEDER OC[C@@H](O)[C@@H](O)C\C(=N\NC(=O)c1ccccc1)\C=N\NC(=O)c2ccccc2
IUPAC-namn N-[(E)-[(2Z,4S,5R)-2-(benzoylhydrazinylidene)-4,5,6-trihydroxyhexylidene]amino]benzamide
Molekylvikt (g/mol) 398.41
Formel vikt 398.159
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.