Läs mer
3-Deoxygalactosone (>90%), TRC
Produktkod. 30136626
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136626 5 mg
30134241 50 mg
30004156 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136626 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2371005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4134-97-8
Molekylformel C6 H10 O5
Molekylvikt (g/mol) 162.14
Synonym D-threo-Hexos-2-ulose, 3-deoxy-, D-threo-Hexosulose, 3-deoxy- (6CI,7CI,8CI), 3-Deoxy-D-threo-hexos-2-ulose, 3-Deoxy-D-galactosone, 3-Deoxygalactosone, (4R,5R)-4,5,6-Trihydroxy-2-oxohexanal
IUPAC-namn (4R,5R)-4,5,6-trihydroxy-2-oxohexanal
LEDER [C@@H](CC(C=O)=O)([C@@H](CO)O)O
Specifikationer
Kemiskt namn eller material 3-Deoxy-galactosone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C6H10O5/c7-2-4(9)1-5(10)6(11)3-8/h2,5-6,8,10-11H,1,3H2/t5-,6-/m1/s1
Formel vikt 162.0528
Procent renhet >95%