Läs mer
3-Deoxyyunaconitine, TRC
Produktkod. 30354718
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354718 10 mg
30361625 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30354718 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28182010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79592-91-9
Molekylformel C35 H49 N O10
Molekylvikt (g/mol) 643.76
Synonym (1α,6α,14α,16β)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)aconitane-8,13,14-triol 8-Acetate 14-(4-methoxybenzoate)
LEDER CCN1C[C@]2(COC)CC[C@H](OC)[C@@]34[C@@H]5C[C@]6(O)[C@H](C[C@](OC(=O)C)([C@H]([C@H](OC)[C@H]23)C14)[C@H]5[C@H]6OC(=O)c7ccc(OC)cc7)OC
Specifikationer
Kemiskt namn eller material 3-Deoxyyunaconitine
Kvantitet 10 mg
InChI formel InChI=1S/C35H49NO10/c1-8-36-17-32(18-40-3)14-13-23(42-5)35-22-15-33(39)24(43-6)16-34(46-19(2)37,26(29(35)36)27(44-7)28(32)35)25(22)30(33)45-31(38)20-9-11-21(41-4)12-10-20/h9-12,22-30,39H,8,13-18H2,1-7H3/t22-,23+,24+,25-,26-,27+,28-,29?,30-,32+,33+,34-,35+/m1/s1
Formel vikt 643.3356