Läs mer
3-Desacetyl cefotaxime lactone, TRC
Produktkod. 30350081
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30350081 5 mg
1 options
Produktkod. 30350081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2715205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (5aR,6R)-6-[[(2Z)-(2-aminothiazol-4-yl)(methoxyimino)acetyl]amino]-5a,6-dihydro-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazine-1,7(4H)-dione
CAS 66340-33-8
Molekylformel C14 H13 N5 O5 S2
Kvantitet 5 mg
Synonym 4-Thiazoleacetamide, 2-amino-α-(methoxyimino)-N-(1,4,5a,6-tetrahydro-1,7-dioxo-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazin-6-yl)-, [5aR-[5aα,6β(Z)]]- (9CI), 3H,7H-Azeto[2,1-b]furo[3,4-d][1,3]thiazine, 4-thiazoleacetamide deriv., Ceftriaxone Sodium Imp. B, Cefotaxime Sodium Imp. E (EP), (5aR,6R)-6-[[(2Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetyl]amino]-5a,6-dihydro-3H,7H-azeto[2,1-b]furo[3,4-d][1,3]thiazine-1,7(4H)-dione, Deacetylcefotaxime lactone
InChI formel InChI=1S/C14H13N5O5S2/c1-23-18-7(6-4-26-14(15)16-6)10(20)17-8-11(21)19-9-5(2-24-13(9)22)3-25-12(8)19/h4,8,12H,2-3H2,1H3,(H2,15,16)(H,17,20)/b18-7-/t8-,12-/m1/s1
LEDER CO\N=C(/C(=O)N[C@H]1[C@H]2SCC3=C(N2C1=O)C(=O)OC3)\c4csc(N)n4
Molekylvikt (g/mol) 395.41
Formel vikt 395.0358