Läs mer
3-Desbromo-3-chloro-Chlorantraniliprole, TRC
Produktkod. 30361634
Change view
Kvantitet:
2.5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361634 2.5 mg
30364240 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361634 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2215602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Desbromo-3-chloro-Chlorantraniliprole
CAS 500008-62-8
Molekylformel C18 H14 Cl3 N5 O2
Kvantitet 2.5 mg
Synonym 3-Chloro-N-(4-chloro-2-methyl-6-(methylcarbamoyl)phenyl)-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxamide
InChI formel InChI=1S/C18H14Cl3N5O2/c1-9-6-10(19)7-11(17(27)22-2)15(9)24-18(28)13-8-14(21)25-26(13)16-12(20)4-3-5-23-16/h3-8H,1-2H3,(H,22,27)(H,24,28)
LEDER CNC(=O)c1cc(Cl)cc(C)c1NC(=O)c2cc(Cl)nn2c3ncccc3Cl
IUPAC-namn 5-chloro-N-[4-chloro-2-methyl-6-(methylcarbamoyl)phenyl]-2-(3-chloropyridin-2-yl)pyrazole-3-carboxamide
Molekylvikt (g/mol) 438.695
Formel vikt 437.021