Läs mer
3-Descyano Fludioxonil 3-Carboxylic Acid, TRC
Produktkod. 30365292
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365292 500 mg
30358204 50 mg
30357828 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289771500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1582788-89-3
Molekylformel C12 H7 F2 N O4
Molekylvikt (g/mol) 267.19
Synonym 1H-Pyrrole-3-carboxylic acid, 4-(2,2-difluoro-1,3-benzodioxol-4-yl)-, 4-(2,2-Difluoro-1,3-benzodioxol-4-yl)-1H-pyrrole-3-carboxylic acid
IUPAC-namn 4-(2,2-difluoro-1,3-benzodioxol-4-yl)-1H-pyrrole-3-carboxylic acid
LEDER OC(=O)c1c[nH]cc1c2cccc3OC(F)(F)Oc23
Specifikationer
Kemiskt namn eller material Fludioxonil-carboxylic acid
Kvantitet 500 mg
InChI formel InChI=1S/C12H7F2NO4/c13-12(14)18-9-3-1-2-6(10(9)19-12)7-4-15-5-8(7)11(16)17/h1-5,15H,(H,16,17)
Formel vikt 267.0343