Läs mer
3-(Dichloroacetyl)-2,2,5-trimethyloxazolidine, TRC
Produktkod. 30180069
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180069 10 mg
30180184 100 mg
30180313 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180069 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD66695510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52836-31-4
Molekylformel C8 H13 Cl2 N O2
Molekylvikt (g/mol) 226.1
Synonym Oxazolidine, 3-(dichloroacetyl)-2,2,5-trimethyl- (9CI), 2,2-Dichloro-1-(2,2,5-trimethyl-3-oxazolidinyl)ethanone, 2,2,5-Trimethyl-3-(dichloroacetyl)-1,3-oxazolidine, 2,2,5-Trimethyl-3-(dichloroacetyl)oxazolidine, R 29148
IUPAC-namn 2,2-dichloro-1-(2,2,5-trimethyl-1,3-oxazolidin-3-yl)ethanone
LEDER CC1CN(C(=O)C(Cl)Cl)C(C)(C)O1
Specifikationer
Kemiskt namn eller material 3-(dichloroacetyl)-2,2,5-trimethyloxazolidine
Kvantitet 10 mg
InChI formel InChI=1S/C8H13Cl2NO2/c1-5-4-11(7(12)6(9)10)8(2,3)13-5/h5-6H,4H2,1-3H3
Formel vikt 225.0323