missing translation for 'onlineSavingsMsg'
Läs mer

3-(Difluoromethoxy)benzylamine, 98%, Thermo Scientific™

Produktkod. 11409490
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11409490 5 g 5g
11499480 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11409490 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H50322.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(Difluoromethoxy)benzylamine is used as pharmaceutical intermediate.

Solubility
Difficult to mix.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 244022-71-7
Molekylformel C8H9F2NO
Molekylvikt (g/mol) 173.163
MDL-nummer MFCD00236230
InChI-nyckel SYAQBBDPVPDVLJ-UHFFFAOYSA-N
Synonym 3-difluoromethoxy benzylamine, 3-difluoromethoxy phenyl methanamine, 3-difluoromethoxy benzyl amine, 1-3-difluoromethoxy phenyl methanamine, benzenemethanamine, 3-difluoromethoxy, benzenemethanamine, 3-difluoromethoxy-9ci, 3-difluoromethoxy phenyl methylamine, acmc-20alqb, 3-difluoromethoxy-benzylamine
PubChem CID 2063350
IUPAC-namn [3-(difluoromethoxy)phenyl]methanamine
LEDER C1=CC(=CC(=C1)OC(F)F)CN

Specifikationer

Kemiskt namn eller material 3-(Difluoromethoxy)benzylamine
Brytningsindex 1.488
Kvantitet 5 g
UN-nummer UN2735
Känslighet Air sensitive
Löslighetsinformation Difficult to mix.
Formel vikt 173.16
Procent renhet 98%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.