Learn More
3-(Difluoromethyl)-1-fluorobenzene, 97%, Thermo Scientific™
Catalog Number p-4812859
Click to view available options
Quantity:
1 g
5 g
26029-52-7
C7H5F3
146.11
MFCD06657983
IKCXLEHVIIUYOA-UHFFFAOYSA-N
3-difluoromethyl-1-fluorobenzene, 1-difluoromethyl-3-fluorobenzene, 3-difluoromethyl fluorobenzene, intermediates-zcf02695, benzene, 1-difluoromethyl-3-fluoro, 1-fluoro-3-difluoromethyl benzene
2761203
1-(difluoromethyl)-3-fluorobenzene
FC(F)C1=CC(F)=CC=C1
26029-52-7
C7H5F3
146.11
MFCD06657983
IKCXLEHVIIUYOA-UHFFFAOYSA-N
3-difluoromethyl-1-fluorobenzene, 1-difluoromethyl-3-fluorobenzene, 3-difluoromethyl fluorobenzene, intermediates-zcf02695, benzene, 1-difluoromethyl-3-fluoro, 1-fluoro-3-difluoromethyl benzene
2761203
1-(difluoromethyl)-3-fluorobenzene
FC(F)C1=CC(F)=CC=C1
Boiling Point 74°C (130.0 Torr)
Flash Point 23°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 146.11
Percent Purity 97%
Chemical Name or Material 3-(Difluoromethyl)-1-fluorobenzene
compliance-icons compliance-icons
Product Identifier
  • 3-(Difluoromethyl)-1-fluorobenzene, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.