missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethyl)-1-fluorobenzene, 97%, ACROS Organics™

Brand:  Acros Organics 451390050

Additional Details : CAS Number : 26029-52-7 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 15209766

  • 1650.79 SEK / 5g


Estimated Shipment
03-11-2020
Add to basket


Specifications

Chemical Identifiers

26029-52-7
146.11
3-difluoromethyl-1-fluorobenzene, 1-difluoromethyl-3-fluorobenzene, 3-difluoromethyl fluorobenzene, intermediates-zcf02695, benzene, 1-difluoromethyl-3-fluoro, 1-fluoro-3-difluoromethyl benzene
1-(difluoromethyl)-3-fluorobenzene
C7H5F3
IKCXLEHVIIUYOA-UHFFFAOYSA-N
2761203
C1=CC(=CC(=C1)F)C(F)F

Specifications

3-(Difluoromethyl)-1-fluorobenzene
74°C (130.0 Torr)
26029-52-7
C7H5F3
IKCXLEHVIIUYOA-UHFFFAOYSA-N
1-(difluoromethyl)-3-fluorobenzene
2761203
97%
23°C
5g
96% min. (GC)
3-difluoromethyl-1-fluorobenzene, 1-difluoromethyl-3-fluorobenzene, 3-difluoromethyl fluorobenzene, intermediates-zcf02695, benzene, 1-difluoromethyl-3-fluoro, 1-fluoro-3-difluoromethyl benzene
C1=CC(=CC(=C1)F)C(F)F
146.11
146.11
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(Difluoromethyl)-1-fluorobenzene, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents