Läs mer
3-Difluoromethyl-1-methylpyrazole-4-carboxylic Acid Ethyl Ester, TRC
Produktkod. 30137558
Change view
Kvantitet:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137558 25 g
30134112 100 g
30006863 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44686025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 141573-95-7
Molekylformel C8H10F2N2O2
Molekylvikt (g/mol) 204.17
Synonym 3-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic Acid Ethyl Ester;1-Methyl-3-(difluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester, Ethyl 1-Methyl-3-(difluoromethyl)pyrazole-4-carboxylate, Ethyl 1-Methyl-3-difluoromethyl-1H-pyrazole-4-carboxylate, Ethyl 3-(Difluoromethyl)-1-methyl-7H-pyrazole-4-carboxylate, Ethyl 3-Difluoromethyl-1-methyl-1H-pyrazole-4-carboxylate
LEDER CCOC(=O)c1cn(C)nc1C(F)F
Specifikationer
Kemiskt namn eller material 3-Difluoromethyl-1-methylpyrazole-4-carboxylic Acid Ethyl Ester
Kvantitet 25 g
InChI formel InChI=1S/C8H10F2N2O2/c1-3-14-8(13)5-4-12(2)11-6(5)7(9)10/h4,7H,3H2,1-2H3
Formel vikt 204.071