Läs mer
3-(Difluoromethyl)-1H-pyrazole-4-carboxylic Acid, TRC
Produktkod. 30138260
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138260 25 mg
30135095 5 mg
30005074 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138260 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44775825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151734-02-0
Molekylformel C5 H4 F2 N2 O2
Molekylvikt (g/mol) 162.09
Synonym Bixafen Metabolite M44, 1H-Pyrazole-4-carboxylic acid, 3-(difluoromethyl)-, 3-(Difluoromethyl)-1H-pyrazole-4-carboxylic acid, 1H-3-Difluoromethylpyrazole-4-carboxylic acid
IUPAC-namn 3-(difluoromethyl)-1H-pyrazole-4-carboxylic acid
LEDER OC(=O)c1c[nH]nc1C(F)F
Specifikationer
Kemiskt namn eller material Bixafen Metabolite M44
Kvantitet 25 mg
InChI formel InChI=1S/C5H4F2N2O2/c6-4(7)3-2(5(10)11)1-8-9-3/h1,4H,(H,8,9)(H,10,11)
Formel vikt 162.0241