missing translation for 'onlineSavingsMsg'
Läs mer

3-Dimethylamino-1-phenylpropan-1-ol, Mikromol

Produktkod. 30459504
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30459504 4 x 25 mg
17982116 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30459504 Supplier Mikromol Supplier No. MM0256.1000254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Dimethylamino-1-phenylpropan-1-ol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Dimethylamino-1-phenylpropan-1-ol
CAS 5554-64-3
Analyt- eller komponentnamn 3-Dimethylamino-1-phenylpropan-1-ol
Frakt skick Room Temperature
Molekylformel C11 H17 N O
Certifikat och efterlevnad ISO 9001
Synonym 3-Dimethylamino-1-phenylpropan-1-ol
InChI formel InChI=1S/C11H17NO/c1-12(2)9-8-11(13)10-6-4-3-5-7-10/h3-7,11,13H,8-9H2,1-2H3
Lösningstyp Neat
LEDER CN(C)CCC(O)c1ccccc1
IUPAC-namn 3-(dimethylamino)-1-phenylpropan-1-ol
Molekylvikt (g/mol) 179.26
Kvantitet 4 x 25 mg
Formel vikt 179.131 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.