missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)benzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number p-4228614
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
23501-93-1
C9H13NO
151.21
MFCD00017342
YTUXZVSDDHCTBZ-UHFFFAOYSA-N
3-dimethylamino phenyl methanol, 3-dimethylamino benzyl alcohol, 3-dimethylaminobenzyl alcohol, m-dimethylamino benzyl alcohol, benzenemethanol,3-dimethylamino, 3-dimethylamino phenyl methan-1-ol, benzyl alcohol, m-dimethylamino, benzenemethanol, 3-dimethylamino
90133
[3-(dimethylamino)phenyl]methanol
CN(C)C1=CC=CC(=C1)CO
This item is not returnable. View return policy
23501-93-1
C9H13NO
151.21
MFCD00017342
YTUXZVSDDHCTBZ-UHFFFAOYSA-N
3-dimethylamino phenyl methanol, 3-dimethylamino benzyl alcohol, 3-dimethylaminobenzyl alcohol, m-dimethylamino benzyl alcohol, benzenemethanol,3-dimethylamino, 3-dimethylamino phenyl methan-1-ol, benzyl alcohol, m-dimethylamino, benzenemethanol, 3-dimethylamino
90133
[3-(dimethylamino)phenyl]methanol
CN(C)C1=CC=CC(=C1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 109.0°C to 110.0°C (0.1 Torr)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 151.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)benzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Dimethylamino)benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'