Läs mer
[[3-[(Dimethylamino)carbonyl]-2-pyridinyl]sulfonyl]carbamic Acid Methyl Ester, TRC
Produktkod. 30372554
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30372554 25 mg
1 options
Produktkod. 30372554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46086625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material ((3-((Dimethylamino)carbonyl)-2-pyridinyl)sulfonyl)carbamic Acid Methyl Ester
CAS 144098-18-0
Molekylformel C10 H13 N3 O5 S
Kvantitet 25 mg
Synonym Methyl N-[3-(N,N-Dimethylcarbamoyl)pyridin-2-ylsulfonyl]carbamate, N-[[3-[(Dimethylamino)carbonyl]-2-pyridinyl]sulfonyl]carbamic Acid Methyl Ester
InChI formel InChI=1S/C10H13N3O5S/c1-13(2)9(14)7-5-4-6-11-8(7)19(16,17)12-10(15)18-3/h4-6H,1-3H3,(H,12,15)
LEDER O=S(C1=NC=CC=C1C(N(C)C)=O)(NC(OC)=O)=O
IUPAC-namn methyl N-[3-(dimethylcarbamoyl)pyridin-2-yl]sulfonylcarbamate
Molekylvikt (g/mol) 287.29
Formel vikt 287.06