Läs mer
3-[(Dimethylamino)carbonyl]phenol, TRC
Produktkod. 30379853
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30379853 250 mg
1 options
Produktkod. 30379853 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD461635250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[(Dimethylamino)carbonyl]phenol
CAS 15789-03-4
Molekylformel C9H11NO2
Kvantitet 250 mg
Synonym 3-Hydroxy-N,N-dimethylbenzamide, m-Hydroxy-N,N-dimethylbenzamide, N,N-Dimethyl-3-hydroxybenzamide
InChI formel InChI=1S/C9H11NO2/c1-10(2)9(12)7-4-3-5-8(11)6-7/h3-6,11H,1-2H3
LEDER CN(C)C(=O)c1cccc(O)c1
Molekylvikt (g/mol) 165.19
Formel vikt 165.079