Läs mer
3-Dimethylaminopropyl-2-benzylaminobenzoate Hydrochloride, TRC
Produktkod. 30132389
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132389 100 mg
17911065 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132389 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD447565100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2196185-65-4
Molekylformel C19 H24 N2 O2 . Cl H
Molekylvikt (g/mol) 348.87
Synonym Benzoic acid, 2-[(phenylmethyl)amino]-, 3-(dimethylamino)propyl ester, hydrochloride (1:1), 3-(Dimethylamino)propyl 2-[(phenylmethyl)amino]benzoate hydrochloride, 3-Dimethylaminopropyl-2-benzylaminobenzoate hydrochloride, 2-[(Phenylmethyl)amino]benzoic acid 3-(Dimethylamino)propyl ester hydrochloride
IUPAC-namn 3-(dimethylamino)propyl 2-(benzylamino)benzoate;hydrochloride
LEDER Cl.CN(C)CCCOC(=O)c1ccccc1NCc2ccccc2
Specifikationer
Kemiskt namn eller material 3-Dimethylaminopropyl 2-benzylaminobenzoate hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C19H24N2O2.ClH/c1-21(2)13-8-14-23-19(22)17-11-6-7-12-18(17)20-15-16-9-4-3-5-10-16;/h3-7,9-12,20H,8,13-15H2,1-2H3;1H
Formel vikt 348.1605
Procent renhet >95%