missing translation for 'onlineSavingsMsg'
Learn More

3-epi-Ochratoxin A, TRC

Product Code. 30410441
Change view
Click to view available options
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30410441 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410441 Supplier Toronto Research Chemicals Supplier No. TRCO1484955MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-epi-Ochratoxin A
CAS 189152-21-4
Molekylformel C20H18ClNO6
Kvantitet 5 mg
Synonym 3S,14S-Ochratoxin A, N-[[(3S)-5-Chloro-3,4-dihydro-8-hydroxy-3-methyl-1-oxo-1H-2-benzopyran-7-yl]carbonyl]-L-phenylalanine
InChI formel InChI=1S/C20H18ClNO6/c1-10-7-12-14(21)9-13(17(23)16(12)20(27)28-10)18(24)22-15(19(25)26)8-11-5-3-2-4-6-11/h2-6,9-10,15,23H,7-8H2,1H3,(H,22,24)(H,25,26)/t10-,15-/m0/s1
LEDER ClC1=C2C(C(O[C@H](C2)C)=O)=C(C(C(N[C@H](C(O)=O)CC3=CC=CC=C3)=O)=C1)O
IUPAC-namn ((S)-5-chloro-8-hydroxy-3-methyl-1-oxoisochromane-7-carbonyl)-L-phenylalanine
Molekylvikt (g/mol) 403.81
Formel vikt 403.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.