Läs mer
3-Ethoxy-1,2-propanediol, TRC
Produktkod. 17964445
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964445 5 g
17982985 1 g
17950985 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8927705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1874-62-0
Molekylformel C5 H12 O3
Molekylvikt (g/mol) 120.147
Synonym Glycerin 1-Ethyl Ether, Glycerol 1-Ethyl Ether, Glycerol α-Ethyl Ether, Glycerol α-Monoethyl Ethe
IUPAC-namn 3-ethoxypropane-1,2-diol
LEDER CCOCC(O)CO
Specifikationer
Kemiskt namn eller material 3-Ethoxy-1,2-propanediol
Anteckningar om renhetsgrad GC
Kvantitet 5 g
InChI formel InChI=1S/C5H12O3/c1-2-8-4-5(7)3-6/h5-7H,2-4H2,1H3
Formel vikt 120.0786
Procent renhet >95%