Läs mer
3-Ethyl-1,5,6,7-tetrahydro-2-methyl-4H-indol-4-one, TRC
Produktkod. 30388485
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30388485 100 mg
30371016 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30388485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE926290100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6116-76-3
Molekylformel C11H15NO
Molekylvikt (g/mol) 177.24
Synonym 2-Methyl-3-ethyl-4-oxo-4,5,6,7-tetrahydroindole, NSC 136938
LEDER CCc1c(C)[nH]c2CCCC(=O)c12
Specifikationer
Kemiskt namn eller material 3-Ethyl-1,5,6,7-tetrahydro-2-methyl-4H-indol-4-one
Kvantitet 100 mg
InChI formel InChI=1S/C11H15NO/c1-3-8-7(2)12-9-5-4-6-10(13)11(8)9/h12H,3-6H2,1-2H3
Formel vikt 177.1154