Learn More
Alfa Aesar™ 3-Ethyl-1-methyl-2-thiohydantoin, 95%
Brand:  Alfa Aesar™ H63948.03
Additional Details : CAS Number : 64895-98-3 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15450939
  • 3114.20 SEK / 1g
Estimated Shipment date
from Supplier 03-09-2018
Add to basket
Specifications
Chemical Identifiers
64895-98-3
158.219
QVLPHASWYNLQSK-UHFFFAOYSA-N
5001929
C6H10N2OS
MFCD00185945
3-Ethyl-1-methyl-2-thioxo-4-imidazolidinone
Specifications
3-Ethyl-1-methyl-2-thiohydantoin
1g
C6H10N2OS
3-Ethyl-1-methyl-2-thioxo-4-imidazolidinone
CCN1C(=O)CN(C1=S)C
158.219
158.22
64895-98-3
95%
MFCD00185945
QVLPHASWYNLQSK-UHFFFAOYSA-N
3-ethyl-1-methyl-2-sulfanylideneimidazolidin-4-one
5001929
Solid