missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methylbenzoxazolium iodide, 95%, Thermo Scientific™

Catalog Number p-7024330
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5260-37-7
C10H12INO
289.12
MFCD00044141
UZWYLWSUBQEUMC-UHFFFAOYSA-M
3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade
21331
3-ethyl-2-methyl-1,3-benzoxazol-3-ium;iodide
[I-].CC[N+]1=C(C)OC2=CC=CC=C12
This item is not returnable. View return policy
5260-37-7
C10H12INO
289.12
MFCD00044141
UZWYLWSUBQEUMC-UHFFFAOYSA-M
3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade
21331
3-ethyl-2-methyl-1,3-benzoxazol-3-ium;iodide
[I-].CC[N+]1=C(C)OC2=CC=CC=C12

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 196°C to 198°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 289.12
Percent Purity 95%
Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'